COM448617598 Combi-Blocks, Inc.

5-Bromo-2-[(3-carboxypropyl)amino]benzoic acid

  • MDL number MFCD22383678
  • Cas Number : 1393441-79-6
  • Molecular Weight : 302.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5397
Availability details for
Quantity
  • Availability
  • Add to Cart