COM448617608 Combi-Blocks, Inc.

4-Bromo-2-(cyanomethyl)benzoic acid

  • MDL number MFCD22383679
  • Cas Number : 942935-70-8
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5414
Availability details for
Quantity
  • Availability
  • Add to Cart