COM448617609 Combi-Blocks, Inc.

4-Bromo-2-methoxy-N,N-dimethylbenzamide

  • MDL number MFCD22383680
  • Cas Number : 1369774-49-1
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5415
Availability details for
Quantity
  • Availability
  • Add to Cart