COM448617610 Combi-Blocks, Inc.

4-Bromo-N,N-diethyl-2-methoxybenzamide

  • MDL number MFCD22383681
  • Cas Number : 1393442-26-6
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5416

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5416
Availability details for
Quantity
  • Availability
  • Add to Cart