COM448617618 Combi-Blocks, Inc.

2-(Propylsulfanyl)benzoic acid

  • MDL number MFCD02729229
  • Cas Number : 21213-10-5
  • Molecular Weight : 196.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5430
Availability details for
Quantity
  • Availability
  • Add to Cart