COM448617619 Combi-Blocks, Inc.

3-Methoxy-5-(trifluoromethoxy)benzoic acid

  • MDL number MFCD18398450
  • Cas Number : 1261442-97-0
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5431
Availability details for
Quantity
  • Availability
  • Add to Cart