COM448617633 Combi-Blocks, Inc.

3-(Benzylamino)-4-nitrobenzoic acid

  • MDL number MFCD22421498
  • Cas Number : 1400644-88-3
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5447
Availability details for
Quantity
  • Availability
  • Add to Cart