COM448617638 Combi-Blocks, Inc.

Methyl 3-(isopropylamino)-4-nitrobenzoate

  • MDL number MFCD22421500
  • Cas Number : 1400645-23-9
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5454
Availability details for
Quantity
  • Availability
  • Add to Cart