COM448617641 Combi-Blocks, Inc.

2,4-Dioxo-4-phenylbutanoic acid

  • MDL number MFCD00456054
  • Cas Number : 5817-92-5
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5462
Availability details for
Quantity
  • Availability
  • Add to Cart