COM448617656 Combi-Blocks, Inc.

Methyl 3-fluoro-4-(trifluoromethyl)benzoate

  • MDL number MFCD06203754
  • Cas Number : 773873-89-5
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5483
Availability details for
Quantity
  • Availability
  • Add to Cart