COM448617667 Combi-Blocks, Inc.

Ethyl 3,5-dichloro-2-iodobenzoate

  • MDL number MFCD22689090
  • Cas Number : 939989-95-4
  • Molecular Weight : 344.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5496
Availability details for
Quantity
  • Availability
  • Add to Cart