COM448617669 Combi-Blocks, Inc.

2-(3-Methoxyphenyl)acetamide

  • MDL number MFCD00085105
  • Cas Number : 18463-71-3
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5499
Availability details for
Quantity
  • Availability
  • Add to Cart