COM448617671 Combi-Blocks, Inc.

3-Methanesulfonyl-4-nitrobenzoic acid

  • MDL number MFCD22586685
  • Cas Number : 1415819-84-9
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5501
Availability details for
Quantity
  • Availability
  • Add to Cart