COM448617676 Combi-Blocks, Inc.

4-tert-Butyl-3,5-dinitrobenzoic acid

  • MDL number MFCD20529168
  • Cas Number : 67688-82-8
  • Molecular Weight : 268.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5508
Availability details for
Quantity
  • Availability
  • Add to Cart