COM448617679 Combi-Blocks, Inc.

(2E)-3,4,4-trimethylpent-2-enamide

  • MDL number MFCD19206375
  • Cas Number : 860546-40-3
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5511
Availability details for
Quantity
  • Availability
  • Add to Cart