COM448617681 Combi-Blocks, Inc.

Sodium (2-cyanophenyl)methanesulfonate

  • MDL number MFCD00024991
  • Cas Number : 197311-76-5
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5513
Availability details for
Quantity
  • Availability
  • Add to Cart