COM448617686 Combi-Blocks, Inc.

4-Amino-2-ethoxy-5-fluorobenzonitrile

  • MDL number MFCD22689093
  • Cas Number : 1420800-17-4
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5519
Availability details for
Quantity
  • Availability
  • Add to Cart