COM448617691 Combi-Blocks, Inc.

Methyl 4-chloro-3-(methylsulfanyl)benzoate

  • MDL number MFCD22489119
  • Cas Number : 166811-70-7
  • Molecular Weight : 216.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5527
Availability details for
Quantity
  • Availability
  • Add to Cart