COM448617703 Combi-Blocks, Inc.

N-(3-Cyanophenyl)cyclopropanecarboxamide

  • MDL number MFCD03159449
  • Cas Number : 101946-36-5
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5544
Availability details for
Quantity
  • Availability
  • Add to Cart