COM448617705 Combi-Blocks, Inc.

(5-Chloro-4-methyl-2-nitrophenyl)acetic acid

  • MDL number MFCD24448778
  • Cas Number : 1437794-56-3
  • Molecular Weight : 229.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5546
Availability details for
Quantity
  • Availability
  • Add to Cart