COM448617708 Combi-Blocks, Inc.

Ethyl 3-nitro-4-(propylamino)benzoate

  • MDL number MFCD02089291
  • Cas Number : 312922-00-2
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5551
Availability details for
Quantity
  • Availability
  • Add to Cart