COM448617711 Combi-Blocks, Inc.

Methyl 5-bromo-2,3-dimethoxybenzoate

  • MDL number MFCD06203939
  • Cas Number : 83131-08-2
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5554
Availability details for
Quantity
  • Availability
  • Add to Cart