COM448617713 Combi-Blocks, Inc.

5-Bromo-2-(cyanomethyl)benzoic acid

  • MDL number MFCD22491974
  • Cas Number : 942935-79-7
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5556
Availability details for
Quantity
  • Availability
  • Add to Cart