COM448617714 Combi-Blocks, Inc.

Dimethyl thiophene-2,3-dicarboxylate

  • MDL number MFCD06203890
  • Cas Number : 14300-68-6
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5560
Availability details for
Quantity
  • Availability
  • Add to Cart