COM448617715 Combi-Blocks, Inc.

4-Chlorophthalic acid

  • MDL number MFCD00044755
  • Cas Number : 89-20-3
  • Molecular Weight : 200.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5561
Availability details for
Quantity
  • Availability
  • Add to Cart