COM448617722 Combi-Blocks, Inc.

2,3-Difluoro-5-iodobenzoic acid

  • MDL number MFCD16875978
  • Cas Number : 333780-74-8
  • Molecular Weight : 284.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5571
Availability details for
Quantity
  • Availability
  • Add to Cart