COM448617728 Combi-Blocks, Inc.

Ethyl 2-(4-ethoxy-4-oxobutanamido)benzoate

  • MDL number MFCD03931427
  • Cas Number : 120572-43-2
  • Molecular Weight : 293.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5577

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5577
Availability details for
Quantity
  • Availability
  • Add to Cart