COM448617731 Combi-Blocks, Inc.

Methyl 5-cyano-2-methoxybenzoate

  • MDL number MFCD11044990
  • Cas Number : 40757-12-8
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5580
Availability details for
Quantity
  • Availability
  • Add to Cart