COM448617740 Combi-Blocks, Inc.

1-(4-Chlorophenyl)cyclopropane-1-carboxamide

  • MDL number MFCD10671794
  • Cas Number : 133284-47-6
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5593
Availability details for
Quantity
  • Availability
  • Add to Cart