COM448617741 Combi-Blocks, Inc.

(2,6-dichloro-3-fluorophenyl)-2-oxoacetic acid

  • MDL number MFCD26131145
  • Cas Number : 1820673-56-0
  • Molecular Weight : 237.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5596
Availability details for
Quantity
  • Availability
  • Add to Cart