COM448617748 Combi-Blocks, Inc.

Ethyl 2,2-difluoro-2-(4-oxochromen-3-yl)acetate

  • MDL number MFCD26383411
  • Cas Number : 1431841-88-1
  • Molecular Weight : 268.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5609
Availability details for
Quantity
  • Availability
  • Add to Cart