COM448617749 Combi-Blocks, Inc.

t-Butyl methyl fumarate

  • MDL number MFCD26383412
  • Cas Number : 55556-66-6
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5610
Availability details for
Quantity
  • Availability
  • Add to Cart