COM448617755 Combi-Blocks, Inc.

3-Bromo-5-fluorobenzamide

  • MDL number MFCD07780673
  • Cas Number : 933585-20-7
  • Molecular Weight : 218.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5618
Availability details for
Quantity
  • Availability
  • Add to Cart