COM448617763 Combi-Blocks, Inc.

Methyl 2,3-dihydro-1,4-benzodioxine-5-carboxylate

  • MDL number MFCD01546256
  • Cas Number : 214894-91-4
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5628

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5628
Availability details for
Quantity
  • Availability
  • Add to Cart