COM448617764 Combi-Blocks, Inc.

2,3-dihydro-1,4-benzodioxine-5-carboxamide

  • MDL number MFCD03659722
  • Cas Number : 349550-81-8
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5629

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5629
Availability details for
Quantity
  • Availability
  • Add to Cart