COM448617774 Combi-Blocks, Inc.

N-BOC 2-Acetyl-4-bromoaniline

  • MDL number MFCD18909632
  • Cas Number : 341521-29-7
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5644

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5644
Availability details for
Quantity
  • Availability
  • Add to Cart