COM448617775 Combi-Blocks, Inc.

(2S)-2-Amino-3-(1,3-benzothiazol-2-yl)propanoic acid hydrochloride

  • MDL number MFCD27935901
  • Cas Number : 1820581-64-3
  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5645
Availability details for
Quantity
  • Availability
  • Add to Cart