COM448617781 Combi-Blocks, Inc.

4-Bromo-3-(methylsulfamoyl)benzoic acid

  • MDL number MFCD10690222
  • Cas Number : 716358-64-4
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5661
Availability details for
Quantity
  • Availability
  • Add to Cart