COM448617799 Combi-Blocks, Inc.

Methyl 1,3-dioxo-2H-isoindole-5-carboxylate

  • MDL number MFCD00455850
  • Cas Number : 56720-83-3
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5693

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5693
Availability details for
Quantity
  • Availability
  • Add to Cart