COM448617806 Combi-Blocks, Inc.

5-Bromo-2-methanesulfonylbenzonitrile

  • MDL number MFCD18378496
  • Cas Number : 1379309-82-6
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5708

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5708
Availability details for
Quantity
  • Availability
  • Add to Cart