COM448617809 Combi-Blocks, Inc.

3-Carbamoyl-4-fluorobenzoic acid

  • MDL number MFCD28101635
  • Cas Number : 1534121-96-4
  • Molecular Weight : 183.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5716

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5716
Availability details for
Quantity
  • Availability
  • Add to Cart