COM448617810 Combi-Blocks, Inc.

Methyl 2,5-dimethanesulfonylbenzoate

  • MDL number MFCD28101636
  • Cas Number : 1820687-93-1
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5717

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5717
Availability details for
Quantity
  • Availability
  • Add to Cart