COM448617815 Combi-Blocks, Inc.

2-(Cyanomethyl)benzene-1,4-dicarboxylic acid

  • MDL number MFCD28122671
  • Cas Number : 1820710-62-0
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5727

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5727
Availability details for
Quantity
  • Availability
  • Add to Cart