COM448617819 Combi-Blocks, Inc.

N,2-Bis(2-bromophenyl)acetamide

  • MDL number MFCD28127182
  • Cas Number : 1820703-74-9
  • Molecular Weight : 369.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5732
Availability details for
Quantity
  • Availability
  • Add to Cart