COM448617825 Combi-Blocks, Inc.

4-Bromo-3-methyl-5-nitrobenzoic acid

  • MDL number MFCD25963262
  • Cas Number : 408533-82-4
  • Molecular Weight : 260.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5739

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5739
Availability details for
Quantity
  • Availability
  • Add to Cart