COM448617826 Combi-Blocks, Inc.

2-Bromo-3-methyl-6-nitrobenzoic acid

  • MDL number MFCD28143928
  • Cas Number : 1807109-55-2
  • Molecular Weight : 260.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5741
Availability details for
Quantity
  • Availability
  • Add to Cart