COM448617828 Combi-Blocks, Inc.

Methyl 4-(piperidine-1-sulfonyl)benzoate

  • MDL number MFCD01107475
  • Cas Number : 577752-97-7
  • Molecular Weight : 283.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5746
Availability details for
Quantity
  • Availability
  • Add to Cart