COM448617832 Combi-Blocks, Inc.

2,4-Dibromo-3-methyl-6-nitrobenzoic acid

  • MDL number MFCD28143929
  • Cas Number : 1820608-82-9
  • Molecular Weight : 338.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5750

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5750
Availability details for
Quantity
  • Availability
  • Add to Cart