COM448617844 Combi-Blocks, Inc.

4-(Benzyloxy)-2-bromobenzonitrile

  • MDL number MFCD28369575
  • Cas Number : 1044067-64-2
  • Molecular Weight : 288.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5772
Availability details for
Quantity
  • Availability
  • Add to Cart