COM448617845 Combi-Blocks, Inc.

Amino[4-(tert-butylsulfanyl)phenyl]acetic acid

  • MDL number MFCD28369576
  • Cas Number : 1336902-75-0
  • Molecular Weight : 239.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5777
Availability details for
Quantity
  • Availability
  • Add to Cart